Learn More
N-Ethylbenzylamine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 343710250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Ethylbenzylamine | |
14321-27-8 | |
100.0 | |
191.0°C to 194.0°C | |
Authentic | |
Glass bottle | |
1.5100 to 1.5130 | |
MFCD00009031 | |
12,102 | |
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine | |
HVAAHUDGWQAAOJ-UHFFFAOYSA-N | |
N-benzylethanamine | |
84352 | |
95% |
95% | |
94.0 | |
0.9000g/mL | |
66°C | |
95% | |
C9H13N | |
C6H5CH2NHC2H5 | |
25g | |
0.9 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ethyl ether and benzene | |
CCNCC1=CC=CC=C1 | |
135.21 | |
135.21 |
Chemical Identifiers
14321-27-8 | |
135.21 | |
HVAAHUDGWQAAOJ-UHFFFAOYSA-N | |
84352 | |
CCNCC1=CC=CC=C1 |
C9H13N | |
MFCD00009031 | |
n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine | |
N-benzylethanamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 238-265-1
TSCA : TSCA
RUO â Research Use Only