Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylbenzisoxazolium tetrafluoroborate, 98+%

$438.69 - $438.69

Chemical Identifiers

Chemical Name or Material N-Ethylbenzisoxazolium tetrafluoroborate
CAS 4611-62-5
Molecular Formula C9H10BF4NO
Formula Weight 234.99
MDL Number MFCD00031785
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2379414

 
alfa aesar™
B2379414
25g Each for $438.69
Description & Specifications

Specifications

Formula Weight 234.99
Chemical Name or Material N-Ethylbenzisoxazolium tetrafluoroborate
Assay ≥98%
Molecular Formula C9H10BF4NO
Melting Point 108° to 110°C
MDL Number MFCD00031785
CAS 4611-62-5
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.