Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylaniline, 97%

Chemical Identifiers

Chemical Name or Material N-Ethylaniline
CAS 103-69-5
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00009025
UN Number UN2272
Beilstein 507468
Merck Index 14,3764
Synonym N-Phenylethylamine; N-Phenylethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA150600F

  SDS
alfa aesar™
A150600F
2500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA15060AP

  SDS
alfa aesar™
A15060AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Health Hazard 2 GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Odor Odorless
TSCA TSCA
Assay 97%
MDL Number MFCD00009025
Beilstein 507468
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-ETHYLANILINE
CAS 103-69-5
RTECS Number BX9780000
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2272
Quantity 2500mL
Merck Index 14,3764
Health Hazard 1 Danger
Synonym N-Phenylethylamine; N-Phenylethylamine
Chemical Name or Material N-Ethylaniline
Boiling Point 204° to 205°C
Flash Point 85°C (185°F)
Melting Point −64°C
Molecular Formula C8H11N

Applications
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).

Solubility
Soluble in water (50g/L).

Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.