missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylaniline, 97%

Manufacturer:  Alfa Aesar™ A15060AP

 View more versions of this product

Catalog No. AAA15060AP



Description

Description

Applications
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).

Solubility
Soluble in water (50g/L).

Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

N-Ethylaniline
Odorless
-64°C
103-69-5
MFCD00009025
507468
ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
CCNC1=CC=CC=C1
121.183
CHEBI:34870
97%
85°C (185°F)
204°C to 205°C
500mL
C8H11N
UN2272
14,3764
OJGMBLNIHDZDGS-UHFFFAOYSA-N
N-ethylaniline
7670
121.18

Chemical Identifiers

103-69-5
121.183
OJGMBLNIHDZDGS-UHFFFAOYSA-N
7670
N-ethylaniline
C8H11N
MFCD00009025
ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
CHEBI:34870
CCNC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.

GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-ETHYLANILINE

RTECSNumber : BX9780000

TSCA : TSCA

Documents