Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylacetamide, 99%

$54.69 - $167.26

Chemical Identifiers

Chemical Name or Material N-Ethylacetamide
CAS 625-50-3
Molecular Formula CH3CONHCH2CH3
Formula Weight 87.12
MDL Number MFCD00009029
Beilstein 969292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1030009

  SDS
alfa aesar™
L1030009
10g Each for $54.69

AAL1030018

  SDS
alfa aesar™
L1030018
50g Each for $167.26
Description & Specifications

Specifications

Formula Weight 87.12
Chemical Name or Material N-Ethylacetamide
MDL Number MFCD00009029
Molecular Formula CH3CONHCH2CH3
Quantity 10g
TSCA TSCA
Beilstein 969292
Boiling Point 206° to 208°C
RTECS Number AB8500000
Flash Point 106°C (222°F)
Melting Point −32°C
Assay 99%
CAS 625-50-3

Applications
N-ethylacetamide is a suitable nylon model for mechanistic studies of dye fading. N-Ethylacetamide was used to investigate propylene carbonate and a mixture of N-methylformamide and N-ethylacetamide by broadband dielectric and mechanical shear spectroscopy. It was used in determination of reaction products of OH-oxidation of N-methylpyrrolidone under atmospheric conditions.

Solubility
Fully misicible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.