missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyl-p-toluidine, TCI America™

Manufacturer:  TCI America E0179500ML

 View more versions of this product

Catalog No. E0179500ML



Specifications

Specifications

N-Ethyl-p-toluidine
Yellow
622-57-1
MFCD00035793
n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
CCNC1=CC=C(C=C1)C
135.21
135.21
≥98.0% (GC,T)
222°C
500mL
C9H13N
2754
AASABFUMCBTXRL-UHFFFAOYSA-N
N-ethyl-4-methylaniline
61164
Clear Liquid at 20°C

Chemical Identifiers

622-57-1
135.21
AASABFUMCBTXRL-UHFFFAOYSA-N
61164
CCNC1=CC=C(C=C1)C
C9H13N
MFCD00035793
n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
N-ethyl-4-methylaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0187&(3)-0193

TSCA : Yes

Documents