missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyl-p-toluenesulfonamide, TCI America™

Manufacturer:  TCI America E067425G

 View more versions of this product

Catalog No. E067425G



Specifications

Specifications

N-Ethyl-p-toluenesulfonamide
White
25g
C9H13NO2S
OHPZPBNDOVQJMH-UHFFFAOYSA-N
N-ethyl-4-methylbenzenesulfonamide
6637
Crystal-Powder at 20°C
208°C
65°C
80-39-7
n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
CCNS(=O)(=O)C1=CC=C(C=C1)C
199.268
199.27
≥98.0% (HPLC,N)

Chemical Identifiers

80-39-7
199.268
n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
N-ethyl-4-methylbenzenesulfonamide
C9H13NO2S
OHPZPBNDOVQJMH-UHFFFAOYSA-N
6637
CCNS(=O)(=O)C1=CC=C(C=C1)C
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1929

TSCA : Yes

Documents