missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyl-o-toluidine, TCI America™

Manufacturer:  TCI America E017725ML

 View more versions of this product

Catalog No. E017725ML



Specifications

Specifications

N-Ethyl-o-toluidine
Yellow
94-68-8
MFCD00026695
n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine
CCNC1=CC=CC=C1C
135.21
135.21
≥98.0% (GC,T)
214°C
25mL
C9H13N
2754
MWOUGPLLVVEUMM-UHFFFAOYSA-N
N-ethyl-2-methylaniline
7201
Clear Liquid at 20°C

Chemical Identifiers

94-68-8
135.21
MWOUGPLLVVEUMM-UHFFFAOYSA-N
7201
CCNC1=CC=CC=C1C
C9H13N
MFCD00026695
n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine
N-ethyl-2-methylaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0187&(3)-0193

TSCA : Yes

Documents