Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-o-toluidine, 98%

Chemical Identifiers

Chemical Name or Material N-Ethyl-o-toluidine
CAS 94-68-8
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00026695
UN Number UN2754
Beilstein 2689722
Synonym 2-Ethylaminotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA11315AE

 
alfa aesar™
A11315AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.21
Chemical Name or Material N-Ethyl-o-toluidine
Assay 98%
Molecular Formula C9H13N
Beilstein 2689722
Quantity 100mL
Health Hazard 1 Danger
TSCA TSCA
MDL Number MFCD00026695
CAS 94-68-8
UN Number UN2754
Boiling Point 218°C
Flash Point 98°C (208°F)
Synonym 2-Ethylaminotoluene
Health Hazard 2 GHS H Statement
H300-H310-H330-H373-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-ETHYLTOLUIDINES
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H373-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.