Please login to your online account to display your discounted pricing

Alfa Aesar™ N'-Ethyl-N,N-dimethylethylenediamine, 98%

Chemical Identifiers

Chemical Name or Material N'-Ethyl-N,N-dimethylethylenediamine
CAS 123-83-1
Molecular Formula CH3CH2NHCH2CH2N(CH3)2
Formula Weight 116.21
MDL Number MFCD00009034
UN Number UN2734
Beilstein 773675
Synonym N,N-Dimethyl-N'-ethylethylenediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0318006

  SDS
alfa aesar™
L0318006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0318014

  SDS
alfa aesar™
L0318014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.21
MDL Number MFCD00009034
Chemical Name or Material N'-Ethyl-N,N-dimethylethylenediamine
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 1 Danger
CAS 123-83-1
Assay 98%
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Quantity 5g
Molecular Formula CH3CH2NHCH2CH2N(CH3)2
Synonym N,N-Dimethyl-N'-ethylethylenediamine
TSCA TSCA
UN Number UN2734
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
Flash Point 23°C (73°F)
Beilstein 773675
Boiling Point 134° to 135°C
Odor Amine-like

Applications
Reatant in the synthesis of tripod amphiphile. N′-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.

Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.