missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide

$86.32 - $86.32

Chemical Identifiers

CAS 912761-62-7
Molecular Formula C13H20N4O
Formula Weight 248.33
MDL Number MFCD08061068
Synonym N-Ethyl-N-methyl-2-(1-piperazinyl)pyridine-3-carboxamide
Chemical Name or Material N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50927MD SDS Alfa Aesar™
H50927MD
250mg Each for $86.32
AAH5092703 SDS Alfa Aesar™
H5092703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

912761-62-7
248.33
N-Ethyl-N-methyl-2-(1-piperazinyl)pyridine-3-carboxamide
C13H20N4O
MFCD08061068
N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide

Specifications

912761-62-7
248.33
250mg
N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide
C13H20N4O
MFCD08061068
N-Ethyl-N-methyl-2-(1-piperazinyl)pyridine-3-carboxamide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning