Please login to your online account to display your discounted pricing

N-Ethyl-n-butylamine 99%, Acros Organics


Quantity 1kg

Manufacturer: thermo fisher scientific  158230010

 View more versions of this product

Catalog No. AC158230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 101.19
Physical Form Liquid
Quantity 1kg
Color Clear colorless to slightly yellow
TSCA TSCA
RTECS Number EO4880000
Density 0.74g/mL
EINECS Number 236-415-0
Beilstein 04, 157
CAS Max % 100.0
Refractive Index 1.4040 to 1.4060 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
CAS Min % 98.5
Name Note 99%
CAS 13360-63-9
Chemical Name or Material N-Ethyl-n-butylamine
Linear Formula CH3(CH2)3NHC2H5
Assay Percent Range 99%
MDL Number MFCD00009428
Molecular Formula C6H15N

Specifications

Chemical Name or Material N-Ethyl-n-butylamine
Name Note 99%
CAS 13360-63-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H15N
Linear Formula CH3(CH2)3NHC2H5
Formula Weight 101.19
MDL Number MFCD00009428
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents