Please login to your online account to display your discounted pricing

N-Ethyl-n-butylamine 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material N-Ethyl-n-butylamine
Name Note 99%
CAS 13360-63-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H15N
Linear Formula CH3(CH2)3NHC2H5
Formula Weight 101.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC158230010

  SDS
thermo fisher scientific
158230010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 101.19
Quantity 1kg
Color Clear colorless to slightly yellow
EINECS Number 236-415-0
TSCA TSCA
Density 0.74g/mL
RTECS Number EO4880000
Beilstein 04, 157
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.4040 to 1.4060 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N-Ethyl-n-butylamine
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H15N
Linear Formula CH3(CH2)3NHC2H5
MDL Number MFCD00009428
CAS 13360-63-9
CAS Min % 98.5
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.