missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-N-(4-methylbenzyl)-2-nitrobenzenesulfonamide, 97%

Chemical Identifiers

CAS 1421262-83-0
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
MDL Number MFCD22683152
InChI Key XPJKNBKPLZFMST-UHFFFAOYSA-N
Synonym n-ethyl-n-4-methylphenyl methyl-2-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-2-nitrobenzenesulfonamide
PubChem CID 97290229
IUPAC Name N-ethyl-N-[(4-methylphenyl)methyl]-2-nitrobenzenesulfonamide
SMILES CCN(CC1=CC=C(C=C1)C)S(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60897MD View Documents Alfa Aesar™
H60897MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6089703 View Documents Alfa Aesar™
H6089703
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421262-83-0
334.39
XPJKNBKPLZFMST-UHFFFAOYSA-N
97290229
CCN(CC1=CC=C(C=C1)C)S(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
C16H18N2O4S
MFCD22683152
n-ethyl-n-4-methylphenyl methyl-2-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-2-nitrobenzenesulfonamide
N-ethyl-N-[(4-methylphenyl)methyl]-2-nitrobenzenesulfonamide

Specifications

250mg
C16H18N2O4S
n-ethyl-n-4-methylphenyl methyl-2-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-2-nitrobenzenesulfonamide
CCN(CC1=CC=C(C=C1)C)S(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
334.39
334.39
N-Ethyl-N-(4-methylbenzyl)-2-nitrobenzenesulfonamide
1421262-83-0
MFCD22683152
XPJKNBKPLZFMST-UHFFFAOYSA-N
N-ethyl-N-[(4-methylphenyl)methyl]-2-nitrobenzenesulfonamide
97290229
97%