Please login to your online account to display your discounted pricing

N-Ethyl-β-alaninenitrile, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  302490250

 View more versions of this product

Catalog No. AC302490250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 98.08
Physical Form Liquid
Quantity 25g
Color Clear colorless to dark yellow-orange
Density 0.8800g/mL
CAS Max % 100.0
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Storage Information May darken during storage
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Chemical Name or Material N-Ethyl-β-alaninenitrile
Molecular Formula C5H10N2
Name Note 98%
MDL Number MFCD00087227
Assay Percent Range 98%
CAS 21539-47-9

Specifications

Chemical Name or Material N-Ethyl-β-alaninenitrile
Name Note 98%
CAS 21539-47-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10N2
Formula Weight 98.08
MDL Number MFCD00087227
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents