missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyl-β-alaninenitrile, 98%, ACROS Organics™

Chemical Identifiers

CAS 21539-47-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C5H10N2
Formula Weight 98.08
MDL Number MFCD00087227
Chemical Name or Material N-Ethyl-β-alaninenitrile, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC302490250 SDS Thermo Fisher Scientific
302490250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21539-47-9
100.0
C5H10N2
MFCD00087227
≥97.5%
97.5
98%
98.08
N-Ethyl-β-alaninenitrile, 98%

Specifications

0.8800g/mL
25g
97.5
Authentic
1.4320 to 1.4340 (20°C, 589nm)
May darken during storage
C5H10N2
MFCD00087227
≥97.5%
Clear colorless to dark yellow-orange
21539-47-9
100.0
Glass bottle
Liquid
98%
98.08
N-Ethyl-β-alaninenitrile, 98%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.