missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-4-methylbenzenesulfonamide, 98%

$62.46 - $188.14

Chemical Identifiers

CAS 80-39-7
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD00048511
InChI Key OHPZPBNDOVQJMH-UHFFFAOYSA-N
Synonym n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
PubChem CID 6637
IUPAC Name N-ethyl-4-methylbenzenesulfonamide
SMILES CCNS(=O)(=O)C1=CC=C(C=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6190522 View Documents Alfa Aesar™
H6190522
100g Each for $62.46
 
AAH6190536 View Documents Alfa Aesar™
H6190536
500g Each for $188.14
 
Specifications

Chemical Identifiers

80-39-7
199.268
OHPZPBNDOVQJMH-UHFFFAOYSA-N
6637
CCNS(=O)(=O)C1=CC=C(C=C1)C
C9H13NO2S
MFCD00048511
n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
N-ethyl-4-methylbenzenesulfonamide

Specifications

208°C (745mmHg)
100g
C9H13NO2S
n-ethyl-p-toluenesulfonamide, santicizer 3, n-tosylethylamine, benzenesulfonamide, n-ethyl-4-methyl, n-ethyl-p-tolylsulfonamide, n-ethyl-4-toluenesulfonamide, p-tolueneethylsulfonamide, ethyl tosylamide, p-toluenesulfonyl-n-ethylamide, n-ethyl-p-methylbenzenesulfonamide
CCNS(=O)(=O)C1=CC=C(C=C1)C
199.268
199.27
N-Ethyl-4-methylbenzenesulfonamide
63°C to 65°C
80-39-7
MFCD00048511
OHPZPBNDOVQJMH-UHFFFAOYSA-N
N-ethyl-4-methylbenzenesulfonamide
6637
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA