missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-4-methoxybenzamide, 97%

$91.30 - $289.92

Chemical Identifiers

CAS 7403-41-0
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD01211881
InChI Key KIOWCVBPJXEJLN-UHFFFAOYSA-N
Synonym benzamide, n-ethyl-4-methoxy, n-ethyl-4-methoxy-benzamide, 4-methoxy-n-ethylbenzamide, n-ethyl 4-methoxyphenyl carboxamide
PubChem CID 81897
IUPAC Name N-ethyl-4-methoxybenzamide
SMILES CCNC(=O)C1=CC=C(C=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59924MD View Documents Alfa Aesar™
H59924MD
250mg Each for $91.30
 
AAH5992403 missing translation for 'viewDocuments' Alfa Aesar™
H5992403
1g Each for $289.92
 
Specifications

Chemical Identifiers

7403-41-0
179.219
KIOWCVBPJXEJLN-UHFFFAOYSA-N
81897
CCNC(=O)C1=CC=C(C=C1)OC
C10H13NO2
MFCD01211881
benzamide, n-ethyl-4-methoxy, n-ethyl-4-methoxy-benzamide, 4-methoxy-n-ethylbenzamide, n-ethyl 4-methoxyphenyl carboxamide
N-ethyl-4-methoxybenzamide

Specifications

250mg
C10H13NO2
benzamide, n-ethyl-4-methoxy, n-ethyl-4-methoxy-benzamide, 4-methoxy-n-ethylbenzamide, n-ethyl 4-methoxyphenyl carboxamide
CCNC(=O)C1=CC=C(C=C1)OC
179.219
179.22
N-Ethyl-4-methoxybenzamide
7403-41-0
MFCD01211881
KIOWCVBPJXEJLN-UHFFFAOYSA-N
N-ethyl-4-methoxybenzamide
81897
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning