Please login to your online account to display your discounted pricing

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Ethyl-3-carbazolecarboxaldehyde
CAS 7570-45-8
CAS Min % 92.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C15H13NO
Formula Weight 223.27
MDL Number MFCD00004963
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166035000

  SDS
thermo fisher scientific
AC166035000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166031000

  SDS
thermo fisher scientific
166031000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166030250

  SDS
thermo fisher scientific
166030250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥92%
Formula Weight 223.27
Physical Form Crystalline Powder or Granules
Purity Grade Technical Grade
Quantity 500g
Color Yellow to ochre
Melting Point 85° to 91°C
CAS Max % 100.0
CAS Min % 92.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 7570-45-8
Assay Percent Range 94%
MDL Number MFCD00004963
Chemical Name or Material N-Ethyl-3-carbazolecarboxaldehyde
Molecular Formula C15H13NO