Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-2-nitroaniline, 98%

Chemical Identifiers

Chemical Name or Material N-Ethyl-2-nitroaniline
CAS 10112-15-9
Formula Weight 166.18
MDL Number MFCD00007091
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5514803

 
alfa aesar™
H5514803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5514806

 
alfa aesar™
H5514806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55148MD

 
alfa aesar™
H55148MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material N-Ethyl-2-nitroaniline
Refractive Index 1.6450
Physical Form Liquid
Assay 98%
Density 1.19g/mL
MDL Number MFCD00007091
CAS 10112-15-9
Boiling Point 168°C (20mmHg)
Color Colorless to dark orange
Quantity 1g