missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-2-nitroaniline, 98%

Manufacturer:  Alfa Aesar™ H5514806

 View more versions of this product

Catalog No. AAH5514806



Specifications

Specifications

N-Ethyl-2-nitroaniline
113°C (235°F)
168°C (20mmHg)
5g
C8H10N2O2
acmc-20ak4v
CCNC1=CC=CC=C1[N+](=O)[O-]
166.18
166.18
98%
1.19 g/mL
1.645
Colorless to Orange
10112-15-9
MFCD00007091
CQIKVOWCSGXCCG-UHFFFAOYSA-N
N-ethyl-2-nitroaniline
82354
Liquid

Chemical Identifiers

10112-15-9
166.18
CQIKVOWCSGXCCG-UHFFFAOYSA-N
82354
CCNC1=CC=CC=C1[N+](=O)[O-]
C8H10N2O2
MFCD00007091
acmc-20ak4v
N-ethyl-2-nitroaniline
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 233-305-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents