Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-2-(4-piperidinyloxy)acetamide, 98%

$333.00 - $333.00

Chemical Identifiers

Chemical Name or Material N-Ethyl-2-(4-piperidinyloxy)acetamide
CAS 521074-81-7
Molecular Formula C9H18N2O2
Formula Weight 186.26
MDL Number MFCD08061052
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5090603

  SDS
alfa aesar™
H5090603
1g Each for $333.00
Description & Specifications

Specifications

Formula Weight 186.26
Chemical Name or Material N-Ethyl-2-(4-piperidinyloxy)acetamide
Assay 98%
Molecular Formula C9H18N2O2
MDL Number MFCD08061052
CAS 521074-81-7
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.