Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-2-(1-piperazinyl)nicotinamide, 99%

$321.72 - $321.72

Chemical Identifiers

Chemical Name or Material N-Ethyl-2-(1-piperazinyl)nicotinamide
Molecular Formula C12H18N4O
Formula Weight 234.3
MDL Number MFCD08061031
Synonym N-Ethyl-2-(1-piperazinyl)pyridine-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5085603

 
alfa aesar™
H5085603
1g Each for $321.72

AAH50856MD

 
alfa aesar™
H50856MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.3
Chemical Name or Material N-Ethyl-2-(1-piperazinyl)nicotinamide
Assay 99%
Molecular Formula C12H18N4O
MDL Number MFCD08061031
Quantity 1g
Synonym N-Ethyl-2-(1-piperazinyl)pyridine-3-carboxamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.