Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-1-naphthylamine, 98%

$89.59 - $1,326.86

Chemical Identifiers

Chemical Name or Material N-Ethyl-1-naphthylamine
CAS 118-44-5
Molecular Formula C12H13N
Formula Weight 171.25
MDL Number MFCD00003880
Beilstein 2207404
Synonym N-(1-Naphthyl)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1950206

 
alfa aesar™
A1950206
5g Each for $89.59

AAA1950222

 
alfa aesar™
A1950222
100g Each for $1,326.86
Description & Specifications

Specifications

Formula Weight 171.25
CAS 118-44-5
Boiling Point 175° to 176°C (15mmHg)
Quantity 5g
Chemical Name or Material N-Ethyl-1-naphthylamine
TSCA TSCA
Assay 98%
Molecular Formula C12H13N
MDL Number MFCD00003880
Beilstein 2207404
Flash Point 128°C (262°F)
Synonym N-(1-Naphthyl)ethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.