Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethyl-1-butylamine, 99%

Chemical Identifiers

Chemical Name or Material N-Ethyl-1-butylamine
CAS 13360-63-9
Molecular Formula C6H15N
Linear Formula CH3(CH2)3NHCH2CH3
Formula Weight 101.19
MDL Number MFCD00009428
UN Number UN2733
Beilstein 1731324
Synonym N-(n-Butyl)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2288130

  SDS
alfa aesar™
B2288130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.19
UN Number UN2733
Health Hazard 1 Danger
Flash Point 12°C (54°F)
Linear Formula CH3(CH2)3NHCH2CH3
TSCA TSCA
Synonym N-(n-Butyl)ethylamine
Health Hazard 2 GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Assay 99%
Beilstein 1731324
MDL Number MFCD00009428
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
RTECS Number EO4880000
Odor Amine-like
Quantity 250g
Molecular Formula C6H15N
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 13360-63-9
Boiling Point 110°C
Chemical Name or Material N-Ethyl-1-butylamine
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.