missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Ethoxycarbonyl)phthalimide, 97%

Manufacturer:  Alfa Aesar™ A1482236

 View more versions of this product

Catalog No. AAA1482236



Description

Description

Applications
N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

N-(Ethoxycarbonyl)phthalimide
500g
C11H9NO4
196340
VRHAQNTWKSVEEC-UHFFFAOYSA-N
ethyl 1,3-dioxoisoindole-2-carboxylate
31187
97%
78°C to 84°C
22509-74-6
MFCD00005893
n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O
219.196
219.2

Chemical Identifiers

22509-74-6
219.196
VRHAQNTWKSVEEC-UHFFFAOYSA-N
31187
CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O
C11H9NO4
MFCD00005893
n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
ethyl 1,3-dioxoisoindole-2-carboxylate
Safety and Handling

Safety and Handling

RTECSNumber : NR3459500

TSCA : TSCA

SDS Documents