missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(Ethoxycarbonyl)nortropinone, 99%

Manufacturer:  Alfa Aesar™ L1384903

 View more versions of this product

Catalog No. AAL1384903



Specifications

Specifications

N-(Ethoxycarbonyl)nortropinone
1g
C10H15NO3
1530944
ANEJUHJDPGTVIO-UHFFFAOYSA-N
ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
401882
99%
152°C to 153°C (11mmHg)
32499-64-2
MFCD00078171
ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
CCOC(=O)N1C2CCC1CC(=O)C2
197.234
197.23

Chemical Identifiers

32499-64-2
197.234
ANEJUHJDPGTVIO-UHFFFAOYSA-N
401882
CCOC(=O)N1C2CCC1CC(=O)C2
C10H15NO3
MFCD00078171
ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Documents