missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC165352500

 View more versions of this product

Catalog No. AC165352500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
Glass bottle
White to Beige
250g
99.0
99% min. (HPLC)
MFCD00006703
02,191; 03,137; 04,223
1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester
GKQLYSROISKDLL-UHFFFAOYSA-N
ethyl 2-ethoxy-2H-quinoline-1-carboxylate
27833
Crystalline Powder
Authentic
125.0°C to 128.0°C (0.1 mmHg)
63.0°C to 66.0°C
16357-59-8
100.0
C14H17NO3
21, V,3, 28
15, 3566
Solubility in water: insoluble. Other solubilities: soluble in ethanol
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
247.29
247.29
99+%

Chemical Identifiers

16357-59-8
247.29
GKQLYSROISKDLL-UHFFFAOYSA-N
27833
CCOC1C=CC2=CC=CC=C2N1C(=O)OCC
C14H17NO3
MFCD00006703
1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester
ethyl 2-ethoxy-2H-quinoline-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 240-418-2

SDS Documents