Please login to your online account to display your discounted pricing

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  165350250

 View more versions of this product

Catalog No. AC165350250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 247.29
Quantity 25g
Color White to beige
Merck Index 15, 3566
Melting Point 63° to 66°C
Fieser 02,191; 03,137; 04,223
CAS Min % 99.0
CAS Max % 100.0
Beilstein 21, V,3, 28
MDL Number MFCD00006703
Health Hazard 1 Warning
Infrared Spectrum Authentic
Chemical Name or Material N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
CAS 16357-59-8
Synonym EEDQ
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C14H17NO3
Health Hazard 2 GHS P Statement Harmful if inhaled. Harmful in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. Causes skin irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
CAS 16357-59-8
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C14H17NO3
Formula Weight 247.29
MDL Number MFCD00006703
Synonym EEDQ
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Harmful if inhaled. Harmful in contact with skin. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. Causes skin irritation.

SDS & Safety Documents