Please login to your online account to display your discounted pricing

Alfa Aesar™ N(epsilon)-Boc-L-lysine, 97%

$354.09 - $354.09

Chemical Identifiers

Chemical Name or Material N(epsilon)-Boc-L-lysine
CAS 2418-95-3
Molecular Formula C11H22N2O4
Formula Weight 246.31
MDL Number MFCD00037221
Beilstein 4252546
Synonym N(epsilon)-tert-Butoxycarbonyl-L-lysine; H-Lys(Boc)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2173806

  SDS
alfa aesar™
B2173806
5g Each for $354.09
Description & Specifications

Specifications

Formula Weight 246.31
Quantity 5g
CAS 2418-95-3
Synonym N(epsilon)-tert-Butoxycarbonyl-L-lysine; H-Lys(Boc)-OH
Molecular Formula C11H22N2O4
MDL Number MFCD00037221
Melting Point ≈250°C (dec.)
Chemical Name or Material N(epsilon)-Boc-L-lysine
Beilstein 4252546
Assay 97%

Applications
It is used as therapy to treat recurrent herpes simplex infections.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.