Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecyltrichlorosilane, 96%

Chemical Identifiers

Chemical Name or Material n-Dodecyltrichlorosilane
CAS 4484-72-4
Molecular Formula C12H25Cl3Si
Linear Formula CH3(CH2)11SiCl3
Formula Weight 303.78
MDL Number MFCD00000483
UN Number UN1771
Beilstein 1758004
Synonym Trichlorododecylsilane; Lauryltrichlorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1317814

  SDS
alfa aesar™
A1317814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1317822

  SDS
alfa aesar™
A1317822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1317836

  SDS
alfa aesar™
A1317836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 303.78
Chemical Name or Material n-Dodecyltrichlorosilane
TSCA TSCA
Assay 96%
Molecular Formula C12H25Cl3Si
Linear Formula CH3(CH2)11SiCl3
MDL Number MFCD00000483
CAS 4484-72-4
UN Number UN1771
Beilstein 1758004
Synonym Trichlorododecylsilane; Lauryltrichlorosilane
Boiling Point 155°C (10mmHg)
Quantity 25g
Flash Point 165°C (329°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DODECYLTRICHLOROSILANE

Applications
n-Dodecyltrichlorosilane is employed in the medicinal field, electronic industry and used as polymer material. It is a coupling and reducing agent. It is also used as a lubricant.

Solubility
Soluble in parts of organic solvents.

Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.