Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecyl thiocyanate, 97%

$42.64 - $310.39

Chemical Identifiers

Chemical Name or Material n-Dodecyl thiocyanate
CAS 765-15-1
Molecular Formula CH3(CH2)11SCN
Formula Weight 227.41
MDL Number MFCD00019773
Beilstein 1770396
Synonym Lauryl thiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0490206

  SDS
alfa aesar™
L0490206
5g Each for $42.64

AAL0490222

  SDS
alfa aesar™
L0490222
100g Each for $310.39
Description & Specifications

Specifications

Formula Weight 227.41
MDL Number MFCD00019773
Assay 97%
CAS 765-15-1
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material n-Dodecyl thiocyanate
Health Hazard 1 Warning
Boiling Point 180°C (18mmHg)
Synonym Lauryl thiocyanate
Beilstein 1770396
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Quantity 5g
RTECS Number XK9625000
Molecular Formula CH3(CH2)11SCN
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.