Please login to your online account to display your discounted pricing

n-Dodecyl Sulfide 93%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material n-Dodecyl sulfide
Name Note 93%
CAS 2469-45-6
CAS Min % 91.0
CAS Max % 100.0
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C24H50S
Linear Formula [CH3(CH2)11]2S
Formula Weight 370.71
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117680250

  SDS
fisher scientific acros
117680250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Flakes
Assay ≥91%
Formula Weight 370.71
Quantity 25g
Color White to light yellow
EINECS Number 219-587-1
TSCA TSCA
Melting Point 36° to 43°C
CAS Max % 100.0
CAS Min % 91.0
Chemical Name or Material n-Dodecyl sulfide
Name Note 93%
Infrared Spectrum Authentic
Assay Percent Range 93%
Molecular Formula C24H50S
Packaging Glass bottle
Linear Formula [CH3(CH2)11]2S
MDL Number MFCD00008975
CAS 2469-45-6
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.