Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecyl gallate, 98%

$50.18 - $163.16

Chemical Identifiers

Chemical Name or Material n-Dodecyl gallate
CAS 1166-52-5
Molecular Formula C19H30O5
Formula Weight 338.45
MDL Number MFCD00002195
Beilstein 2701981
Synonym n-Dodecyl 3,4,5-trihydroxybenzoate; Gallic acid n-dodecyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0623318

  SDS
alfa aesar™
L0623318
50g Each for $50.18

AAL0623330

  SDS
alfa aesar™
L0623330
250g Each for $163.16
Description & Specifications

Specifications

Formula Weight 338.45
Chemical Name or Material n-Dodecyl gallate
MDL Number MFCD00002195
CAS 1166-52-5
Molecular Formula C19H30O5
Melting Point 95° to 98°C
Beilstein 2701981
Quantity 50g
Synonym n-Dodecyl 3,4,5-trihydroxybenzoate; Gallic acid n-dodecyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DH9100000
TSCA TSCA
Assay 98%
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.