Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecyl gallate, 98%


Quantity 50g

Manufacturer: alfa aesar™  L0623318

 View more versions of this product

Catalog No. AAL0623318

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 338.45
Chemical Name or Material n-Dodecyl gallate
MDL Number MFCD00002195
CAS 1166-52-5
Molecular Formula C19H30O5
Melting Point 95° to 98°C
Beilstein 2701981
Quantity 50g
Synonym n-Dodecyl 3,4,5-trihydroxybenzoate; Gallic acid n-dodecyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DH9100000
TSCA TSCA
Assay 98%
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.

Specifications

Chemical Name or Material n-Dodecyl gallate
CAS 1166-52-5
Molecular Formula C19H30O5
Formula Weight 338.45
MDL Number MFCD00002195
Beilstein 2701981
Synonym n-Dodecyl 3,4,5-trihydroxybenzoate; Gallic acid n-dodecyl ester
Accessories

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

SDS & Safety Documents