missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Dodecyl-β-D-maltoside, 99%, high purity, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 329370010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Dodecyl-β-D-maltoside | |
98.5 | |
217.0°C | |
Authentic | |
Glass bottle | |
MFCD00043012 | |
17, V,8, 12 | |
+ 47.50 | |
Solubility in water: soluble (100g/L). Other solubilities: soluble in methanol, ethanol | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
510.6 | |
CHEBI:43769 | |
99% | |
Powder |
69227-93-6 | |
100.0 | |
White | |
98.5% min. (ELSD) (HPLC) | |
C24H46O11 | |
1 g | |
+47.50 (20.00°C c=1,water) | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
114880 | |
510.6 | |
High Purity |
Chemical Identifiers
69227-93-6 | |
510.6 | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
114880 | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C24H46O11 | |
MFCD00043012 | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
CHEBI:43769 | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
RUO – Research Use Only