Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecyl 4-hydroxybenzoate, 98%

$227.64 - $227.64

Chemical Identifiers

Chemical Name or Material n-Dodecyl 4-hydroxybenzoate
CAS 2664-60-0
Molecular Formula C19H30O3
Formula Weight 306.45g/mol
MDL Number MFCD00016473
Beilstein 2739355
Synonym 4-Hydroxybenzoic acid n-dodecyl ester; Lauryl 4-hydroxybenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0629214

 
alfa aesar™
L06292-14
25g Each for $227.64
Description & Specifications

Specifications

Melting Point 54° to 56°C
Quantity 25g
Chemical Name or Material n-Dodecyl 4-hydroxybenzoate
CAS 2664-60-0
Assay 98%
Molecular Formula C19H30O3
Formula Weight 306.45g/mol
MDL Number MFCD00016473
Beilstein 2739355
Synonym 4-Hydroxybenzoic acid n-dodecyl ester; Lauryl 4-hydroxybenzoate
RTECS Number DH2185600