Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecanethiol gold nanoparticles, 2% (w/v) soln. in toluene, 2-5nm particles

Chemical Identifiers

Chemical Name or Material n-Dodecanethiol gold nanoparticles
Name Note 2 to 5nm particles
MDL Number MFCD09953506
UN Number UN1294
Synonym Gold-n-dodecanethiol in toluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH29045AA

  SDS
alfa aesar™
H29045AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material n-Dodecanethiol gold nanoparticles
Name Note 2 to 5nm particles
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H304-H361-H373-H315-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
Concentration 2% (w/v) soln. in toluene
MDL Number MFCD09953506
UN Number UN1294
Quantity 5mL
Synonym Gold-n-dodecanethiol in toluene
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H304-H361-H373-H315-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.