missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Dodecane, ≥99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A148340F

 View more versions of this product

Catalog No. AAA148340F



Description

Description

n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

Solubility
Miscible with ethyl alcohol, ethyl ether, acetone, chloroform and carbon tetrachloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Dodecane
-10°C
215°C to 217°C
≥99%
1.422
2500 mL
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
SNRUBQQJIBEYMU-UHFFFAOYSA-N
dodecane
8182
170.34
112-40-3
0.749
71°C (159°F)
C12H26
MFCD00008969
1697175
Miscible with ethyl alcohol,ethyl ether,acetone,chloroform and carbon tetrachloride. Immiscible with water.
CCCCCCCCCCCC
170.34
CHEBI:28817
Liquid

Chemical Identifiers

112-40-3
170.34
SNRUBQQJIBEYMU-UHFFFAOYSA-N
8182
dodecane
C12H26
MFCD00008969
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
CHEBI:28817
CCCCCCCCCCCC
Safety and Handling

Safety and Handling

P210-P280-P301+P310-P331-P370+P378q-P501c

H227-H304

EINECSNumber : 203-967-9

RTECSNumber : JR2125000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only