Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Dodecane, 99%

$28.32 - $28.32

Chemical Identifiers

Chemical Name or Material n-Dodecane
CAS 112-40-3
Linear Formula CH3(CH2)10CH3
Formula Weight 170.34
MDL Number MFCD00008969
Beilstein 1697175
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3132414

  SDS
alfa aesar™
3132414
25g Each for $28.32
Description & Specifications

Specifications

Chemical Name or Material n-Dodecane
CAS 112-40-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number JR2125000
TSCA TSCA
Refractive Index 1.4220
Physical Form Liquid
Assay 99%
Density 0.749g/mL
Linear Formula CH3(CH2)10CH3
Formula Weight 170.34
Melting Point −10°C
MDL Number MFCD00008969
Boiling Point 215° to 217°C
Beilstein 1697175
Color Colorless
Quantity 25g
Flash Point 71° (159°F)
Odor Petrol-like
Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.