missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Docosane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158785000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Docosane | |
98.5 | |
43.0°C to 46.0°C | |
0.7700g/mL | |
>112°C | |
98.5% min. (GC) | |
C22H46 | |
MFCD00009348 | |
01, 174 | |
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane | |
HOWGUJZVBDQJKV-UHFFFAOYSA-N | |
docosane | |
12405 | |
310.6 | |
Crystalline Solid |
629-97-0 | |
100.0 | |
White | |
369.0°C | |
Authentic | |
Glass bottle | |
CH3(CH2)20CH3 | |
500 g | |
0.77 | |
Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene | |
CCCCCCCCCCCCCCCCCCCCCC | |
310.61 | |
CHEBI:46050 | |
99% |
Chemical Identifiers
629-97-0 | |
310.61 | |
HOWGUJZVBDQJKV-UHFFFAOYSA-N | |
12405 | |
docosane |
C22H46 | |
MFCD00009348 | |
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane | |
CHEBI:46050 | |
CCCCCCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : 211-121-5
RUO – Research Use Only