missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Docosane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 158780250

 View more versions of this product

Catalog No. AC158780250



Description

Description

Specifications

Specifications

n-Docosane
>112°C
Glass bottle
369.0°C
43.0°C to 46.0°C
629-97-0
100.0
C22H46
MFCD00009348
ch3-ch2 20-ch3, docosane, docosane, analytical standard, docosane, n, dokosan, heneicosane, methyl, n-docosane, normal-docosane, twt, unii-ow99q363ko
HOWGUJZVBDQJKV-UHFFFAOYSA-N
docosane
12405
310.6
99%
0.7700g/mL
Authentic
0.77
White
25g
98.5
98.5% min. (GC)
CH3(CH2)20CH3
01, 174
Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene
CCCCCCCCCCCCCCCCCCCCCC
310.61
CHEBI:46050
Crystalline Solid

Chemical Identifiers

629-97-0
310.61
HOWGUJZVBDQJKV-UHFFFAOYSA-N
12405
docosane
C22H46
MFCD00009348
ch3-ch2 20-ch3, docosane, docosane, analytical standard, docosane, n, dokosan, heneicosane, methyl, n-docosane, normal-docosane, twt, unii-ow99q363ko
CHEBI:46050
CCCCCCCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

EINECSNumber : 211-121-5

SDS Documents