Please login to your online account to display your discounted pricing

N-Dimethoxymethyl-N,N-dimethylamine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB00604EA

 View more versions of this product

Catalog No. SB00604EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 119.16
Quantity 10g
Assay 97%
Density 0.8900g/mL
CAS Min % 97.0
Packaging Amber Glass Bottle
Linear Formula (CH3)2NCH(OCH3)2
Synonym 1, 1-Dimethoxytrimethylamine
CAS 149-73-5
CAS 68-12-2
CAS 67-56-1
MDL Number MFCD00008482
CAS 4637-24-5
Health Hazard 2 GHS H Statement:
May damage the unborn child.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material N-Dimethoxymethyl-N, N-dimethylamine
Assay Percent Range 97%
Health Hazard 1 Danger
Molecular Formula C5H13NO2

Specifications

Chemical Name or Material N-Dimethoxymethyl-N, N-dimethylamine
CAS 4637-24-5
CAS Min % 97.0
CAS 149-73-5
CAS 68-12-2
CAS 67-56-1
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H13NO2
Linear Formula (CH3)2NCH(OCH3)2
Formula Weight 119.16
MDL Number MFCD00008482
Synonym 1, 1-Dimethoxytrimethylamine
Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child.
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents