Learn More
n-Decylamine, 99%, Thermo Scientific Chemicals
$80.79 - $280.66
Chemical Identifiers
CAS | 2016-57-1 |
---|---|
Molecular Formula | C10H23N |
Molecular Weight (g/mol) | 157.30 |
MDL Number | MFCD00008149 |
InChI Key | MHZGKXUYDGKKIU-UHFFFAOYSA-N |
Synonym | decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine |
PubChem CID | 8916 |
IUPAC Name | decan-1-amine |
SMILES | CCCCCCCCCCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC111920100
|
Thermo Scientific Chemicals
111920100 |
10 g | Glass bottle |
Each for $80.79
|
|
||||
AC111920500
|
Thermo Scientific Chemicals
111920500 |
50 g | Glass bottle |
Each for $280.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2016-57-1 | |
157.30 | |
MHZGKXUYDGKKIU-UHFFFAOYSA-N | |
8916 | |
CCCCCCCCCCN |
C10H23N | |
MFCD00008149 | |
decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine | |
decan-1-amine |
Specifications
2016-57-1 | |
100.0 | |
Brown or Colorless to Yellow | |
216°C to 218°C | |
Authentic | |
Glass bottle | |
1.4350 to 1.437 | |
MFCD00008149 | |
04, 199 | |
decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine | |
CCCCCCCCCCN | |
157.30 | |
157.3 | |
Liquid |
98.5 | |
12°C to 15°C | |
0.7870g/mL | |
85°C | |
98.5% min. (GC) | |
C10H23N | |
CH3(CH2)9NH2 | |
10 g | |
0.787 | |
MHZGKXUYDGKKIU-UHFFFAOYSA-N | |
decan-1-amine | |
8916 | |
99% | |
n-Decylamine |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT i
GHS Signal Word: Danger
EINECSNumber : 217-957-7
RUO – Research Use Only