Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Decyl methyl sulfide, 97%

$69.46 - $256.15

Chemical Identifiers

Chemical Name or Material n-Decyl methyl sulfide
CAS 22438-39-7
Molecular Formula C11H24S
Linear Formula CH3(CH2)9SCH3
Formula Weight 188.37
MDL Number MFCD00026541
Beilstein 1736530
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0547318

  SDS
alfa aesar™
L0547318
50g Each for $256.15

AAL0547309

  SDS
alfa aesar™
L0547309
10g Each for $69.46
Description & Specifications

Specifications

Formula Weight 188.37
Chemical Name or Material n-Decyl methyl sulfide
Assay 97%
Molecular Formula C11H24S
MDL Number MFCD00026541
CAS 22438-39-7
Beilstein 1736530
Quantity 50g
Flash Point 101°C (214°F)
Linear Formula CH3(CH2)9SCH3
Boiling Point 126° to 127°C (14mmHg)