missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Decanoyl-N-methyl-D-glucamine

Non-ionic detergent for solubilizing membrane proteins

$117.05 - $441.04

Chemical Identifiers

CAS 85261-20-7
Molecular Formula C17H35NO6
Molecular Weight (g/mol) 349.468
MDL Number MFCD00036801
InChI Key UMWKZHPREXJQGR-XOSAIJSUSA-N
Synonym n-oxodecyl meglumine, unii-9q27za5m8g, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, decanoyl n-methylglucamide, decanoylmethylglucamine, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino-, stereoisomer
PubChem CID 174432
IUPAC Name N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
SMILES CCCCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6017303 View Documents Alfa Aesar™
J6017303
1g Each for $117.05
 
AAJ6017306 View Documents Alfa Aesar™
J6017306
5g Each for $441.04
 
Specifications

Chemical Identifiers

85261-20-7
349.468
UMWKZHPREXJQGR-XOSAIJSUSA-N
174432
CCCCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O
C17H35NO6
MFCD00036801
n-oxodecyl meglumine, unii-9q27za5m8g, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, decanoyl n-methylglucamide, decanoylmethylglucamine, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino-, stereoisomer
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide

Specifications

White
1g
C17H35NO6
6974359
Soluble in methanol at 50mg/ml; Also soluble in water at 10mg/ml 20 °C
CCCCCCCCCC(=O)N(C)CC(C(C(C(CO)O)O)O)O
349.468
349.46
N-Decanoyl-N-methyl-D-glucamine
91°C to 93°C
85261-20-7
MFCD00036801
n-oxodecyl meglumine, unii-9q27za5m8g, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, n-methyl-n-2s,3r,4r,5r-2,3,4,5,6-pentahydroxyhexyl decanamide, decanoyl n-methylglucamide, decanoylmethylglucamine, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino, d-glucitol, 1-deoxy-1-methyl 1-oxodecyl amino-, stereoisomer
UMWKZHPREXJQGR-XOSAIJSUSA-N
N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]decanamide
174432
Powder
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C