Please login to your online account to display your discounted pricing

n-Decanophenone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  299210050

 View more versions of this product

Catalog No. AC299210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 34° to 38°C
Quantity 5g
Chemical Name or Material n-Decanophenone
CAS 6048-82-4
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline solid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H24O
Formula Weight 232.37g/mol
MDL Number MFCD00009582

Specifications

Chemical Name or Material n-Decanophenone
CAS 6048-82-4
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H24O
Formula Weight 232.37g/mol
MDL Number MFCD00009582