missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Decane, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A14732AE

 View more versions of this product

Catalog No. AAA14732AE



Description

Description

Applications
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

Solubility
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

n-Decane
100mL
99%
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
CCCCCCCCCC
142.286
CHEBI:41808
Liquid
0.730
124-18-5
C10H22
DIOQZVSQGTUSAI-UHFFFAOYSA-N
decane
15600
142.29

Chemical Identifiers

124-18-5
142.286
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
CHEBI:41808
CCCCCCCCCC
C10H22
DIOQZVSQGTUSAI-UHFFFAOYSA-N
15600
decane
SDS Documents