missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclopropyl-4-fluorobenzylamine, 97%

Manufacturer:  Alfa Aesar™ H56206MD

 View more versions of this product

Catalog No. AAH56206MD



Description

Description

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-Cyclopropyl-4-fluorobenzylamine
625437-46-9
MFCD07330007
n-4-fluorobenzyl cyclopropanamine, cyclopropyl-4-fluoro-benzyl-amine, n-4-fluorophenyl methyl cyclopropanamine, cyclopropyl-4-fluorobenzyl amine, benzenemethanamine, n-cyclopropyl-4-fluoro, n-cyclopropyl-p-fluorobenzylamine, acmc-209n4r, n-cyclopropyl-4-fluorobenzylamine, n-4-fluorobenzyl cyclopropanamine #, n-4-fluorophenyl methyl cyclopropylamine
C1CC1NCC2=CC=C(C=C2)F
165.211
165.21
250mg
C10H12FN
UN2735
YKNFXOCMGURJMK-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]cyclopropanamine
573031
97%

Chemical Identifiers

625437-46-9
165.211
YKNFXOCMGURJMK-UHFFFAOYSA-N
573031
C1CC1NCC2=CC=C(C=C2)F
C10H12FN
MFCD07330007
n-4-fluorobenzyl cyclopropanamine, cyclopropyl-4-fluoro-benzyl-amine, n-4-fluorophenyl methyl cyclopropanamine, cyclopropyl-4-fluorobenzyl amine, benzenemethanamine, n-cyclopropyl-4-fluoro, n-cyclopropyl-p-fluorobenzylamine, acmc-209n4r, n-cyclopropyl-4-fluorobenzylamine, n-4-fluorobenzyl cyclopropanamine #, n-4-fluorophenyl methyl cyclopropylamine
N-[(4-fluorophenyl)methyl]cyclopropanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

SDS Documents