missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Cyclopropyl-4-fluorobenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5620603

 View more versions of this product

Catalog No. AAH5620603



Description

Description

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-Cyclopropyl-4-fluorobenzylamine
625437-46-9
MFCD07330007
acmc-209n4r
C1CC1NCC2=CC=C(C=C2)F
165.211
165.21
1g
C10H12FN
UN2735
YKNFXOCMGURJMK-UHFFFAOYSA-N
N-[(4-fluorophenyl)methyl]cyclopropanamine
573031
97%

Chemical Identifiers

625437-46-9
165.211
YKNFXOCMGURJMK-UHFFFAOYSA-N
573031
C1CC1NCC2=CC=C(C=C2)F
C10H12FN
MFCD07330007
acmc-209n4r
N-[(4-fluorophenyl)methyl]cyclopropanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents